Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chuk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162410.1 | NP_001155882.1 | 8 | 801 | ||
NM_007700.2 | NP_031726.2 | 8 | 801 | |||
GenBank mRNA | AK035326.1 | - | 8 | 758 | ||
AK150039.1 | - | 8 | 780 | |||
AK155328.1 | - | 8 | 781 | |||
AK155393.1 | - | 8 | 802 | |||
AK164236.1 | - | 8 | 802 | |||
BC018243.1 | - | 8 | 793 | 4511275 | ||
U12473.1 | - | 8 | 769 |