Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007721.4 | NP_031747.2 | 2 | 112 | ||
GenBank mRNA | AF208238.1 | - | 2 | 92 | ||
AF215982.1 | - | 2 | 99 | |||
AF215983.1 | - | 2 | 88 | |||
AK090044.1 | - | 2 | 115 | |||
BC117043.1 | - | 2 | 115 | 40126362 | ||
BC120542.1 | - | 2 | 115 | 40129465 |