Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007750.2 | NP_031776.1 | 2 | 487 | ||
GenBank mRNA | AK002218.1 | - | 2 | 447 | ||
AK002524.1 | - | 2 | 403 | |||
AK008594.1 | - | 2 | 438 | |||
AK187874.1 | - | 1 | 86 | |||
U37721.1 | - | 2 | 408 |