Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284507.1 | NP_001271436.1 | 1 | 197 | ||
NM_013496.3 | NP_038524.1 | 1 | 197 | |||
GenBank mRNA | AK045283.1 | - | 1 | 139 | ||
AK144397.1 | - | 1 | 149 | |||
BC065787.1 | - | 1 | 125 | 891192 | ||
X15481.1 | - | 1 | 123 | |||
X15789.1 | - | 1 | 139 |