Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020891.1 | - | 3 | 181 | ||
BC085185.1 | - | 4 | 323 | 30679384 | ||
M60559.1 | - | 2 | 148 |