Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007774.3 | NP_031800.1 | 3 | 541 | ||
GenBank mRNA | BC056430.1 | - | 3 | 528 | 6824353 | |
CF728682.1 | - | 3 | 541 | |||
CF738355.1 | - | 3 | 541 |