Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005880.3 | NP_005871.1 | 9 | 1272 | ||
GenBank mRNA | AF088046.1 | - | 3 | 248 | ||
AJ001309.1 | - | 9 | 1182 | |||
AK313031.1 | - | 9 | 1220 | |||
BC013044.2 | - | 9 | 1224 | 3505399 | ||
BC015809.2 | - | 9 | 1225 | 3922477 | ||
Y13350.1 | - | 7 | 867 |