Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301284.1 | NP_001288213.1 | 5 | 545 | ||
NM_007806.3 | NP_031832.2 | 6 | 629 | |||
GenBank mRNA | AK018713.1 | - | 5 | 514 | ||
AK021200.1 | - | 6 | 617 | |||
AK078854.1 | - | 6 | 585 | |||
AK131580.1 | - | 6 | 600 | |||
AK152414.1 | - | 6 | 620 | |||
AK171748.1 | - | 6 | 598 | |||
AK190653.1 | - | 1 | 77 | |||
BC026791.1 | - | 6 | 580 | 3980538 | ||
BC060635.1 | - | 5 | 4631 | |||
CF738398.1 | - | 6 | 612 | |||
EU791539.1 | - | 6 | 559 | |||
M31775.1 | - | 6 | 552 |