Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cycs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007808.4 | NP_031834.1 | 3 | 880 | ||
GenBank mRNA | AK002732.1 | - | 3 | 882 | ||
AK003168.1 | - | 3 | 909 | |||
AK143887.1 | - | 3 | 881 | |||
BC034363.1 | - | 3 | 864 | 2650742 | ||
BC094054.1 | - | 3 | 869 | 2865963 |