Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104525.1 | NP_001097995.1 | 3 | 495 | ||
GenBank mRNA | BC145630.1 | - | 3 | 525 |
Gene Symbol: Cyp2c40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010004.2 | NP_034134.2 | 3 | 525 | ||
GenBank mRNA | AF047727.1 | - | 3 | 439 | ||
BC139471.1 | - | 3 | 525 | 8862493 | ||
BC139472.1 | - | 3 | 525 | 8862494 |
Gene Symbol: Cyp2c67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024719.2 | NP_001019890.1 | 3 | 445 | ||
GenBank mRNA | AK149389.1 | - | 3 | 456 | ||
BC092260.1 | - | 3 | 445 | 5132591 | ||
BC106142.1 | - | 3 | 445 | 5051946 |