Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007851.2 | NP_031877.2 | 2 | 273 | ||
GenBank mRNA | AV067547.1 | - | 102 | |||
AV070515.1 | - | 2 | 163 | |||
BC119610.2 | - | 2 | 222 | |||
U03033.1 | - | 1 | 56 |
Gene Symbol: Gm10104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177481.1 | NP_001170952.1 | 2 | 256 |
Gene Symbol: Gm14850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177522.1 | NP_001170993.1 | 256 |
Gene Symbol: Gm15315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177528.1 | NP_001170999.1 | 2 | 228 |
Gene Symbol: Gm21498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270613.1 | NP_001257542.1 | 2 | 256 | ||
GenBank mRNA | AA871410.1 | - | 267 |