Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCKDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122957.2 | NP_001116429.1 | 2 | 514 | ||
NM_001271926.1 | NP_001258855.1 | 2 | 514 | |||
NM_005881.3 | NP_005872.2 | 2 | 514 | |||
XM_017022859.1 | XP_016878348.1 | 2 | 427 | |||
GenBank mRNA | AF026548.1 | - | 2 | 424 | ||
AK130145.1 | - | 2 | 394 | |||
AK294691.1 | - | 1 | 223 | |||
BC007363.2 | - | 2 | 397 | 3630050 | ||
BC009872.2 | - | 2 | 377 | 3944953 | ||
BX427330.2 | - | 2 | 513 | |||
CR542093.1 | - | 1 | 151 |