Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291105.1 | NP_001278034.1 | 7 | 1358 | ||
NM_007891.5 | NP_031917.1 | 7 | 1402 | |||
GenBank mRNA | AF483514.1 | - | 7 | 1244 | ||
AF483515.1 | - | 7 | 1244 | |||
AK017841.1 | - | 7 | 1350 | |||
AK153968.1 | - | 7 | 1367 | |||
AK220226.1 | - | 7 | 1099 | |||
BC052160.1 | - | 7 | 1344 | 6330425 | ||
L21973.1 | - | 7 | 1362 |