Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290711.1 | NP_001277640.1 | 8, 9 | 977 | ||
NM_007897.3 | NP_031923.1 | 8, 9 | 858 | |||
GenBank mRNA | AK035781.1 | - | 8, 9 | 741 | ||
AK137389.1 | - | 8, 9 | 899 | |||
BC139362.1 | - | 8, 9 | 819 | 8862384 | ||
BC139364.1 | - | 8, 9 | 819 | 8862386 | ||
BC139765.1 | - | 1, 2 | 310 | |||
L12147.1 | - | 8, 9 | 824 |