Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017405.2 | NP_001017405.1 | 7 | 873 | ||
NM_001297430.1 | NP_001284359.1 | 6 | 669 | |||
NM_001297431.1 | NP_001284360.1 | 5 | 673 | |||
NM_001297432.1 | NP_001284361.1 | 7 | 1011 | |||
NM_001297433.1 | NP_001284362.1 | 7 | 947 | |||
NM_005882.4 | NP_005873.2 | 6 | 750 | |||
NR_123716.1 | 5 | 673 | ||||
XM_006713849.3 | XP_006713912.1 | 7 | 833 | |||
XM_006713850.2 | XP_006713913.1 | 7 | 1006 | |||
XM_011513376.1 | XP_011511678.1 | 8 | 993 | |||
XM_011513377.1 | XP_011511679.1 | 7 | 911 | |||
XM_017007636.1 | XP_016863125.1 | 8 | 993 | |||
XM_017007637.1 | XP_016863126.1 | 7 | 910 | |||
GenBank mRNA | AB208907.1 | - | 2 | 3573 | ||
AF084928.1 | - | 7 | 711 | |||
AK001088.1 | - | 7 | 833 | |||
AK022515.1 | - | 5 | 642 | |||
AK022586.1 | - | 6 | 710 | |||
AK091951.1 | - | 2 | 744 | |||
AK094253.1 | - | 4 | 494 | |||
AK127875.1 | - | 3 | 1101 | |||
AK128302.1 | - | 2 | 350 | |||
AK296174.1 | - | 6 | 626 | |||
AK298944.1 | - | 7 | 927 | |||
AK300572.1 | - | 8 | 770 | |||
AK301155.1 | - | 7 | 841 | |||
AY189687.1 | - | 7 | 786 | |||
AY236486.1 | - | 5 | 618 | |||
BC001225.1 | - | 7 | 840 | 3533990 | ||
BC006470.1 | - | 7 | 796 | |||
BT006957.1 | - | 7 | 777 |