Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007916.2 | NP_031942.2 | 2, 3 | 206 | ||
GenBank mRNA | AK049467.1 | - | 2, 3 | 177 | ||
AK146548.1 | - | 2, 3 | 167 | |||
BC011270.1 | - | 2, 3 | 178 | 3674608 | ||
BY302446.1 | - | 2, 3 | 206 | |||
L25125.1 | - | 2, 3 | 203 |