Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017405.2 | NP_001017405.1 | 8, 9 | 1158 | ||
NM_001297430.1 | NP_001284359.1 | 7, 8 | 954 | |||
NM_001297431.1 | NP_001284360.1 | 6, 7 | 954 | |||
NM_001297432.1 | NP_001284361.1 | 8, 9 | 1296 | |||
NM_001297433.1 | NP_001284362.1 | 8, 9 | 1232 | |||
NM_005882.4 | NP_005873.2 | 7, 8 | 1035 | |||
NR_123716.1 | 6, 7 | 958 | ||||
XM_017007636.1 | XP_016863125.1 | 9, 10 | 1278 | |||
XM_017007637.1 | XP_016863126.1 | 8, 9 | 1195 | |||
GenBank mRNA | AF084928.1 | - | 8, 9 | 995 | ||
AK001088.1 | - | 8, 9 | 1118 | |||
AK022515.1 | - | 6, 7 | 923 | |||
AK022586.1 | - | 7, 8 | 995 | |||
AK091951.1 | - | 3, 4 | 1029 | |||
AK094253.1 | - | 5, 6 | 779 | |||
AK127875.1 | - | 4, 5 | 1386 | |||
AK128302.1 | - | 2, 3 | 2062 | |||
AK296174.1 | - | 7, 8 | 911 | |||
AK298944.1 | - | 8, 9 | 1212 | |||
AK300572.1 | - | 9, 10 | 1055 | |||
AK301155.1 | - | 8, 9 | 1126 | |||
AY189687.1 | - | 8, 9 | 1071 | |||
AY236486.1 | - | 6, 7 | 903 | |||
BC001225.1 | - | 8, 9 | 1125 | 3533990 | ||
BC006470.1 | - | 8, 9 | 1081 | |||
BT006957.1 | - | 8, 9 | 1062 |