Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fau | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160239.2 | NP_001153711.1 | 5 | 466 | ||
NM_001190436.1 | NP_001177365.1 | 4 | 631 | |||
NM_007990.3 | NP_032016.1 | 5 | 469 | |||
GenBank mRNA | AK002355.1 | - | 5 | 407 | ||
AK008466.1 | - | 4 | 376 | |||
AK132752.1 | - | 5 | 412 | |||
AK146359.1 | - | 5 | 404 | |||
AK151211.1 | - | 5 | 407 | |||
AK151465.1 | - | 5 | 407 | |||
AK152738.1 | - | 5 | 407 | |||
AK168348.1 | - | 5 | 407 | |||
AV502224.1 | - | 3 | 232 | |||
BC058691.1 | - | 5 | 386 | 6814998 | ||
BC062873.1 | - | 5 | 398 | 5683472 | ||
BC081463.1 | - | 5 | 386 | 6408436 | ||
D26610.1 | - | 5 | 406 | |||
X65922.1 | - | 5 | 394 |