Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007991.3 | NP_032017.2 | 6, 7 | 755 | ||
GenBank mRNA | AK012620.1 | - | 6, 7 | 751 | ||
AK146328.1 | - | 6, 7 | 747 | |||
AV486126.1 | - | 1, 2 | 65 | |||
BC003813.1 | - | 6, 7 | 767 | 3487295 | ||
BC092274.1 | - | 6, 7 | 793 | 4196645 | ||
BU511282.1 | - | 6, 7 | 762 | |||
Z22593.1 | - | 6, 7 | 743 |