Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010213.3 | NP_034343.2 | 3 | 433 | ||
GenBank mRNA | AF114382.1 | - | 3 | 404 | ||
AF149826.1 | - | 2 | 231 | |||
BC144903.1 | - | 3 | 397 | 9053451 | ||
BC145939.1 | - | 3 | 397 | 40131242 | ||
CB205184.1 | - | 1 | 90 |