Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008066.3 | NP_032092.1 | 9 | 1159 | ||
GenBank mRNA | AK039055.1 | - | 9 | 1160 | ||
AK046953.1 | - | 4 | 1293 | |||
AK048165.1 | - | 1057 | ||||
BC115727.1 | - | 9 | 1081 | 40039253 | ||
M86567.1 | - | 9 | 910 |