Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010253.3 | NP_034383.1 | 4 | 299 | ||
GenBank mRNA | AK133522.1 | - | 4 | 315 | ||
BC044055.1 | - | 4 | 299 | 6476459 | ||
Z23069.1 | - | 2 | 78 |