Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008092.3 | NP_032118.2 | 6 | 1722 | ||
GenBank mRNA | AB075549.1 | - | 6 | 1740 | ||
AF179424.1 | - | 5 | 1143 | |||
AK134639.1 | - | 5 | 746 | |||
BC137824.1 | - | 5 | 1475 | 8860843 | ||
BG088871.2 | - | 1 | 29 | |||
M98339.1 | - | 6 | 1690 | |||
U85046.1 | - | 6 | 1693 |