Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010259.2 | NP_034389.2 | 8 | 1369 | ||
GenBank mRNA | AK077796.1 | - | 8 | 1371 | ||
BC108990.2 | - | 1414 | 40047769 | |||
EF494422.1 | - | 8 | 1369 | |||
M55544.1 | - | 1353 | ||||
M63961.1 | - | 1447 |