Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010267.3 | NP_034397.1 | 6 | 1113 | ||
GenBank mRNA | AK039016.1 | - | 1 | 3081 | ||
AK045460.1 | - | 6 | 1113 | |||
AK083814.1 | - | 6 | 1114 | |||
AK139057.1 | - | 6 | 956 | |||
AK149281.1 | - | 6 | 1124 | |||
BC048177.1 | - | 6 | 1089 | 6825756 | ||
BC051135.1 | - | 6 | 1078 | 6532839 | ||
BC060212.1 | - | 6 | 1087 | |||
Y17850.1 | - | 6 | 1101 |