Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF002701.1 | - | 3, 4 | 432 | ||
AF079107.1 | - | 2, 3 | 445 | |||
AK138512.1 | - | 3, 4 | 1158 | |||
AK149349.1 | - | 3, 4 | 1162 | |||
AK154579.1 | - | 3, 4 | 1168 | |||
BC138561.1 | - | 3, 4 | 553 | 8861581 |