Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010299.3 | NP_034429.1 | 2 | 316 | ||
GenBank mRNA | AK149642.1 | - | 2 | 208 | ||
AK152256.1 | - | 2 | 208 | |||
AK179394.1 | - | 2 | 124 | |||
AK181597.1 | - | 2 | 124 | |||
BC004651.1 | - | 2 | 184 | 3482848 | ||
CT010289.1 | - | 2 | 137 | |||
L19526.1 | - | 2 | 150 | |||
U09816.1 | - | 2 | 193 |