Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spsb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK036451.1 | - | 2 | 399 | ||
AK148177.1 | - | 2 | 955 | |||
BC002005.1 | - | 3 | 1043 | 3496382 | ||
BC010305.1 | - | 1 | 1166 | 3597711 |