Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301340.1 | NP_001288269.1 | 2 | 194 | ||
NM_001301341.1 | NP_001288270.1 | 4 | 321 | |||
NM_001301343.1 | NP_001288272.1 | 2 | 180 | |||
NM_001301344.1 | NP_001288273.1 | 2 | 194 | |||
NM_001301345.1 | NP_001288274.1 | 2 | 194 | |||
NM_025279.3 | NP_079555.1 | 2 | 194 | |||
GenBank mRNA | AK011428.1 | - | 2 | 199 | ||
AK028023.1 | - | 2 | 128 | |||
AK051313.1 | - | 2 | 188 | |||
AK078777.1 | - | 4 | 322 | |||
AK088462.1 | - | 2 | 181 | |||
AK128891.1 | - | 2 | 187 | |||
AK135519.1 | - | 2 | 108 | |||
AK144245.1 | - | 7404 | ||||
AK150848.1 | - | 2 | 174 | |||
AK151689.1 | - | 2 | 148 | |||
AK152927.1 | - | 2 | 148 | |||
AK160615.1 | - | 2 | 164 | |||
AK160887.1 | - | 2 | 196 | |||
AK166656.1 | - | 2 | 199 | |||
AK166927.1 | - | 2 | 152 | |||
AK167600.1 | - | 2 | 185 | |||
AK169064.1 | - | 2 | 184 | |||
BC006694.1 | - | 2 | 161 | 3583037 | ||
BC089328.1 | - | 2 | 117 | 6402614 | ||
BY794631.1 | - | 2 | 181 | |||
EU604547.1 | - | 1 | 47 |