Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008272.3 | NP_032298.1 | 3 | 1610 | ||
GenBank mRNA | AK012173.1 | - | 2 | 998 | ||
AK078758.1 | - | 2 | 1036 | |||
BC050838.1 | - | 3 | 1607 | 6510837 | ||
X55318.1 | - | 2 | 1327 |