Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265588.1 | NP_001252517.1 | 5 | 883 | ||
NM_003414.5 | NP_003405.3 | 4 | 732 | |||
NR_049749.1 | 5 | 776 | ||||
XM_017022865.1 | XP_016878354.1 | 5 | 819 | |||
XM_017022866.1 | XP_016878355.1 | 6 | 803 | |||
GenBank mRNA | AF220492.1 | - | 4 | 685 | ||
AK096390.1 | - | 4 | 593 | |||
AK292573.1 | - | 4 | 557 | |||
AL832123.1 | - | 1 | 2919 | |||
BC036367.1 | - | 4 | 656 | 4828269 | ||
BC127088.1 | - | 4 | 655 | 40123276 | ||
BC127089.1 | - | 4 | 655 | 40123277 | ||
X78925.1 | - | 3 | 490 |