Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038698.1 | NP_001033787.1 | 7 | 1371 | ||
NM_001163397.1 | NP_001156869.1 | 6 | 1126 | |||
NM_001163399.1 | NP_001156871.1 | 7 | 1415 | |||
NM_010488.4 | NP_034618.2 | 7 | 1457 | |||
GenBank mRNA | AF041341.1 | - | 6 | 1138 | ||
AK014133.1 | - | 7 | 1183 | |||
AK079088.1 | - | 7 | 1362 | |||
BC048159.1 | - | 7 | 1430 | 5703005 | ||
BC051987.1 | - | 1128 | ||||
BC052451.1 | - | 7 | 1334 | 6409658 | ||
D31953.1 | - | 7 | 1204 |