Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111320.1 | NP_001104790.1 | 8 | 1335 | ||
NM_010497.3 | NP_034627.3 | 8 | 1313 | |||
GenBank mRNA | AF020039.1 | - | 7 | 1163 | ||
AK087063.1 | - | 8 | 1215 | |||
AK149019.1 | - | 8 | 1244 | |||
AK151212.1 | - | 8 | 1238 | |||
AK159173.1 | - | 8 | 1244 | |||
AK160896.1 | - | 8 | 1245 | |||
AK167158.1 | - | 8 | 1285 | |||
AK167583.1 | - | 8 | 1249 | |||
AK168832.1 | - | 8 | 1286 | |||
BC088986.1 | - | 8 | 1285 | 6808731 | ||
DQ403075.1 | - | 7 | 1063 |