Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008334.3 | NP_032360.2 | 1 | 457 | ||
GenBank mRNA | BC130231.1 | - | 1 | 458 | 40141930 |
Gene Symbol: Ifna11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC116870.1 | - | 1 | 494 | 40126189 | |
BC116872.1 | - | 1 | 494 | 40126191 |