Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008355.3 | NP_032381.1 | 1 | 159 | ||
GenBank mRNA | M23504.1 | - | 1 | 160 |