Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021147.4 | NP_066970.3 | 3 | 1389 | ||
NR_125346.1 | 3 | 1765 | ||||
NR_125347.1 | 3 | 1394 | ||||
NR_125348.1 | 2 | 1259 | ||||
GenBank mRNA | AK026075.1 | - | 3 | 1352 | ||
BC004877.2 | - | 3 | 1645 | |||
X52486.1 | - | 2 | 1146 |