Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lalba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010679.1 | NP_034809.1 | 1 | 119 | ||
GenBank mRNA | BC006922.1 | - | 1 | 131 | 3601746 | |
BC069916.1 | - | 1 | 119 | 3670536 | ||
M80909.1 | - | 1 | 119 |