Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lats1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010690.1 | NP_034820.1 | 4 | 2093 | ||
GenBank mRNA | AF104414.1 | - | 1 | 1218 | ||
AK038612.1 | - | 4 | 2094 | |||
BC158092.1 | - | 3 | 1939 | 9088177 | ||
BC158123.1 | - | 4 | 2088 | 9088196 |