Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCIRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006019.3 | NP_006010.2 | 13 | 1614 | ||
NM_006053.3 | NP_006044.1 | 8 | 1373 | |||
XM_005273709.3 | XP_005273766.1 | 14 | 1773 | |||
XM_011544726.2 | XP_011543028.1 | 13 | 1513 | |||
XM_017017089.1 | XP_016872578.1 | 13 | 1609 | |||
XM_017017090.1 | XP_016872579.1 | 12 | 1952 | |||
GenBank mRNA | AF025374.1 | - | 8 | 1373 | ||
AY548968.1 | - | 1 | 235 | |||
AY548969.1 | - | 2 | 113 | |||
BC018133.1 | - | 13 | 1593 | 3905446 | ||
BC032465.1 | - | 13 | 1586 | 5210733 | ||
U45285.1 | - | 13 | 1542 |