Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141813.1 | - | 13 | 1636 | ||
AK170166.1 | - | 13 | 1962 | |||
AK172083.1 | - | 12 | 1395 | |||
BC063768.1 | - | 13 | 1616 | |||
U14166.1 | - | 13 | 1659 | |||
U15159.1 | - | 13 | 1631 |