Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028796.1 | - | 7 | 1550 | ||
AK156499.1 | - | 5 | 1509 | |||
AK157297.1 | - | 6 | 1419 | |||
AK165124.1 | - | 4 | 739 | |||
AK171128.1 | - | 7 | 1493 | |||
AK172573.1 | - | 6 | 1423 | |||
AK187693.1 | - | 19 | ||||
AK188457.1 | - | 1 | 19 | |||
BC006759.1 | - | 6 | 1403 | 3709861 | ||
U89992.2 | - | 1314 |