Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010734.2 | NP_034864.2 | 1, 2 | 91 | ||
GenBank mRNA | AI182980.1 | - | 1, 2 | 91 | ||
BC086881.1 | - | 1, 2 | 85 | 6747449 | ||
M18187.1 | - | 1, 2 | 84 | |||
U72643.1 | - | 1, 2 | 81 |