Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265589.1 | NP_001252518.1 | 6 | 3106 | ||
NM_001265590.1 | NP_001252519.1 | 5 | 2770 | |||
NM_006054.3 | NP_006045.1 | 4 | 718 | |||
NM_201428.2 | NP_958831.1 | 5 | 3049 | |||
NM_201429.2 | NP_958832.1 | 5 | 775 | |||
NM_201430.2 | NP_958833.1 | 4 | 718 | |||
NR_049750.1 | 3 | 579 | ||||
NR_049751.1 | 3 | 579 | ||||
XM_011544730.2 | XP_011543032.1 | 6 | 3196 | |||
XM_011544731.2 | XP_011543033.1 | 5 | 910 | |||
GenBank mRNA | AB209771.1 | - | 4 | 655 | ||
AF059524.1 | - | 4 | 655 | |||
AF119297.1 | - | 4 | 667 | |||
AK075412.1 | - | 4 | 668 | |||
AK094965.1 | - | 4 | 1771 | |||
AK122727.1 | - | 4 | 1171 | |||
AK127079.1 | - | 5 | 2745 | |||
AK222898.1 | - | 4 | 655 | |||
AK295361.1 | - | 5 | 680 | |||
AK295534.1 | - | 4 | 572 | |||
AK295758.1 | - | 5 | 2698 | |||
AK297533.1 | - | 3 | 529 | |||
AK304100.1 | - | 6 | 3056 | |||
AK307811.1 | - | 5 | 2995 | |||
AK308083.1 | - | 4 | 663 | |||
AK309766.1 | - | 5 | 2724 | |||
AY427821.2 | - | 5 | 3016 | |||
AY750848.1 | - | 6 | 3073 | |||
BC000634.1 | - | 4 | 626 | |||
BC010556.1 | - | 4 | 631 | |||
BC011394.1 | - | 4 | 667 | |||
BC022993.1 | - | 4 | 657 | |||
BC100822.2 | - | 4 | 648 | |||
BC100823.2 | - | 4 | 648 | |||
BC105981.1 | - | 4 | 599 | |||
BC105982.1 | - | 4 | 630 | |||
BC118550.1 | - | 4 | 648 | |||
BC118628.1 | - | 4 | 648 |