Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130796.4 | NP_570614.2 | 2 | 1896 | ||
GenBank mRNA | AF408408.1 | - | 2 | 1884 | ||
AK052343.1 | - | 2 | 1857 | |||
AK076119.1 | - | 2 | 724 | |||
AK077890.1 | - | 2 | 1490 | |||
BC040407.1 | - | 2 | 599 | |||
BC063089.1 | - | 2 | 1896 | 6516923 | ||
BC137801.1 | - | 2 | 1882 | 8860820 | ||
BC145951.1 | - | 2 | 1882 | 40131254 |