Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtap16-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183296.1 | NP_899119.1 | 1 | 372 | ||
GenBank mRNA | AF345293.1 | - | 1 | 312 | ||
AK017265.1 | - | 1 | 372 | |||
BC107303.2 | - | 1 | 308 | |||
BC107304.2 | - | 1 | 308 | |||
BC119503.1 | - | 1 | 155 | |||
BC119504.2 | - | 1 | 308 | |||
BC127231.1 | - | 1 | 308 | |||
BC127232.1 | - | 1 | 308 | |||
BC132658.1 | - | 1 | 350 | 40130935 | ||
BC132660.1 | - | 1 | 350 | 40130937 |