Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265589.1 | NP_001252518.1 | 5 | 3024 | ||
NM_001265590.1 | NP_001252519.1 | 4 | 2688 | |||
NM_006054.3 | NP_006045.1 | 3 | 636 | |||
NM_201428.2 | NP_958831.1 | 4 | 2967 | |||
NM_201429.2 | NP_958832.1 | 4 | 693 | |||
NM_201430.2 | NP_958833.1 | 3 | 636 | |||
XM_011544730.2 | XP_011543032.1 | 5 | 3114 | |||
XM_011544731.2 | XP_011543033.1 | 4 | 828 | |||
GenBank mRNA | AB209771.1 | - | 3 | 573 | ||
AF119297.1 | - | 3 | 585 | |||
AK075412.1 | - | 3 | 586 | |||
AK094965.1 | - | 3 | 1689 | |||
AK122727.1 | - | 3 | 1089 | |||
AK127079.1 | - | 4 | 2663 | |||
AK222898.1 | - | 3 | 573 | |||
AK295361.1 | - | 4 | 598 | |||
AK295534.1 | - | 3 | 490 | |||
AK295758.1 | - | 4 | 2616 | |||
AK304100.1 | - | 5 | 2974 | |||
AK307811.1 | - | 4 | 2913 | |||
AK308083.1 | - | 3 | 581 | |||
AK309766.1 | - | 4 | 2642 | |||
AY427821.2 | - | 4 | 2934 | |||
AY750848.1 | - | 5 | 2991 | |||
BC000634.1 | - | 3 | 544 | |||
BC010556.1 | - | 3 | 549 | |||
BC011394.1 | - | 3 | 585 | |||
BC022993.1 | - | 3 | 575 | |||
BC100822.2 | - | 3 | 566 | |||
BC100823.2 | - | 3 | 566 | |||
BC105981.1 | - | 3 | 517 | |||
BC105982.1 | - | 3 | 548 | |||
BC118550.1 | - | 3 | 566 | |||
BC118628.1 | - | 3 | 566 |