Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285920.1 | NP_001272849.1 | 14 | 1863 | ||
NM_001285921.1 | NP_001272850.1 | 14 | 1846 | |||
NM_001285922.1 | NP_001272851.1 | 13 | 1777 | |||
NM_001285923.1 | NP_001272852.1 | 13 | 1777 | |||
NM_133201.3 | NP_573464.2 | 13 | 1780 | |||
GenBank mRNA | AB048831.1 | - | 11 | 1381 | ||
AF384100.1 | - | 14 | 1660 | |||
AK042080.1 | - | 13 | 1561 | |||
AK049583.1 | - | 13 | 1570 | |||
AK052689.1 | - | 13 | 1561 | |||
AK122220.1 | - | 12 | 2755 | |||
AK146801.1 | - | 14 | 1623 | |||
AK147863.1 | - | 13 | 1576 | |||
AK169373.1 | - | 13 | 1565 | |||
AY028170.1 | - | 1541 | ||||
AY123975.1 | - | 13 | 1727 | |||
BC046503.1 | - | 13 | 1558 | 4191179 |