Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013825.3 | NP_038853.2 | 5 | 818 | ||
GenBank mRNA | AF395445.1 | - | 5 | 818 | ||
AK049301.1 | - | 5 | 861 | |||
AK157921.1 | - | 5 | 882 | |||
BC150734.1 | - | 839 | 9087645 | |||
U19271.1 | - | 852 |