Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17rc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134159.4 | NP_598920.3 | 7, 8 | 914 | ||
NM_178942.1 | NP_849273.1 | 8, 9 | 986 | |||
GenBank mRNA | AF458066.1 | - | 7, 8 | 795 | ||
AK075634.1 | - | 4, 5 | 404 | |||
BC004759.1 | - | 8, 9 | 934 | 3154616 |