Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008540.2 | NP_032566.2 | 12 | 3000 | ||
GenBank mRNA | AK004804.1 | - | 11 | 2365 | ||
AK170255.1 | - | 1 | 481 | |||
BC046584.1 | - | 12 | 3000 | 6313280 |